Tisk…
Cleanroom mops, single-use, King CR
Dodavatel: PFENNIG REINIGUNGSTECHNIK
3500055
141-1362EA 157 CZK
141-1362
Cleanroom mops, single-use, King CR
Mopy
Cleanroom mops with high quality and suitable processing. Made up of 100% polyester.
  • Cost-effective single-use mop
  • Perfect for cytosstatica areas
  • With polyester pocket and high area performance through backfilling
Informace pro objednávání: Optional sterile double-packaged is also available on request.
Balení: Pre-washed and single double packaged.