Tisk…
Klercide™ 70/30 Alcohols, Sterile
Dodavatel: ECOLAB CONTAMINATION CONTROL
3078260 3078300 6600942
141-1190EA 14530 CZK
141-1190 141-1188 SHIE6600942
Klercide™ 70/30 Alcohols, Sterile
Prostředky čistící Dezinfekční prostředek
Contains 70% IPA or denatured ethanol (IMS) blended with deionised water, filtered to 0,2 µm, filled and double bagged in an ISO Class 7; Grade C (microbial count) cleanroom before irradiation using a validated process at no less than 25 kGy to ensure spore free alcohol.
  • SteriShield Delivery System™ ensures 6 months in-use shelf life
  • Certificates of analysis, irradiation and sterility with every batch
  • Fully validated and CEN tested
  • Ideal for Grade A and B cleanrooms and product contact areas
Certifikace: EN standards: EN 1276 - bactericidal activity,More Product Information