Tisk…
HYGEN™ Charging and Wring Buckets
Dodavatel: Rubbermaid Commercial Products
FGQ90088YEL FGQ95088YEL
141-0133EA 6100 CZK
141-0133 129-0940
HYGEN™ Charging and Wring Buckets
Kbelíky
Hygen™ microfibre buckets complete a highly efficient, flat mop cleaning system for healthcare and sanitary maintenance applications.
  • Smooth, non-porous surface helps prevent bacterial growth and cleans up easily
  • Ergonomic, no bending or pulling
  • Press sieve provides easy, no-touch wringing
Rubbermaid Hygen™ microfibre bucket: Mops wick just enough liquid for damp cleaning to ensure quick drying times. Bucket design prevents over-saturation of microfibre mops and features a measuring guide for filling with cleaning solution before mops are added.