Tisk…
Hygen™ microfibre dust mop
Dodavatel: Rubbermaid Commercial Products
R050648
141-0117EA 653 CZK
141-0117
Hygen™ microfibre dust mop
Mopy
The Rubbermaid Commercial HYGEN™ Microfibre Dust Pads trap and lock away dust, dirt, and debris mechanically and electrostatically with split-blended fibres.
  • Dense 3 - 5 micrometer quality Fibre removes more dust, dirt and microbes than conventional cleaning products
  • Advanced hook-and-loop backing holds-up under harsh laundering conditions and attaches pads more securely
  • Double finished edges retain mop shape with use over time
  • Durable for up to 500 launderings (up to 200 with chlorine)
  • Chlorine safe in hot and cold water
For use with Hygen™ PULSE™ mops.