Tisk…
Stěrka pro čisté prostory, NovaSqueegee™
Dodavatel: MICRONOVA
QDNS-12 QDNS-18 QDNS-22 NS-22 RS-12 RS-18 RS-22
129-0531EA 6160 CZK
129-0531 129-0532 129-0533 141-0095 141-0096 141-0097 141-0098
Stěrka pro čisté prostory, NovaSqueegee™
Mopy
A stainless steel squeegee tool with a replaceable rubber blade, designed to clean windows, screens, floors, and walls in the cleanroom. Optional PVA blades are fast-drying, low-linting, and low in particulates with a 600% absorbency rate; ideal for electronic and semiconductor cleanrooms.
  • Designed to clean windows, screens, floors, and walls in the cleanroom
  • Made with 304 electropolished stainless steel and Derlin® resin
  • Rubber replacement blades available
  • PVA replacement blades available for absorbency
  • Quick disconnect handle attachment
  • Entire unit is
More Product Information