Tisk…

Potassium stannate trihydrate ≥95% (metals basis)

Dodavatel: Thermo Fisher Scientific

14015.30 14015.A1
14015.30EA 1350 CZK
14015.30 14015.A1
Potassium stannate trihydrate ≥95% (metals basis)
Potassium stannate trihydrate

Purity: 95+% CAS No.: 12125-03-0 Molecular Formula: K2SnO3.3H2O Formula Weight: 298.94 (244.89anhy) Physical Form: Crystalline Odour: Odourless MDL No.: MFCD00150391

Vzorec: K₂O₃Sn·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150391
CAS číslo: 12125-03-0
EINECS: 235-255-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR