Tisk…

Platinum disulphide (≥74.8% Pt)

Dodavatel: Thermo Fisher Scientific

13992.03 13992.06
13992.03EA 9610 CZK
13992.03 13992.06
Platinum disulphide (≥74.8% Pt)
Platinum disulphide

CAS No.: 12038-21-0 Molecular Formula: PtS2 Formula Weight: 259.22 Physical Form: Powder Melting Point: 225-250° dec. MDL No.: MFCD00016293

Vzorec: PtS₂
Storage Temperature: Ambient
MDL Number: MFCD00016293
CAS číslo: 12038-21-0
EINECS: 234-876-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR