Tisk…
Stem castors for shelving systems, MetroMax iQ
Dodavatel: Metro International
5MDBXA 5MDXA 5PCBX 5PCX 5MPBX 5MPX 5MPXGSA 5PCBXM 5PCXM 5MPBXGSA 5MX 5MBX
139-0045EA 1890 CZK
139-0045 139-0801 139-0802 139-0803 139-0046 139-0047 139-0048 139-0049 139-0050 139-0083 METO5MX METO5MBX
Stem castors for shelving systems, MetroMax iQ
Kolečka
For use with either MetroMax i polymer or MetroMax Q steel stem castor posts.
  • Promotes easy cleaning
  • Castors allow shelving units to be moved easily for access to floors and wall
  • Efficient transport: create a cart to efficiently move supplies throughout a facility or campus
Polymer stem castors:
Corrosion resistant performance: all-polymer horn, stainless steel axle and hardware, 127 mm diameter polyurethane wheel, sleeve-style axle bearing, and an optional toe-operated brakemechanism
Models available with antimicrobial product protection built intoMore Product Information