Tisk…
Utility carts, SP series
Dodavatel: Metro International
2SPN55ABR
139-0773EA 17960 CZK
139-0773
Utility carts, SP series
Vozíky
These carts are designed to transport heavy loads. They consist of Super Erecta wire shelves with plastic split sleeves, two one-piece handles with matching finish and designated castors with donut bumpers. Made from steel, Super Erecta brite finish.
  • Two-tier model
  • 5M resilient rubber castors, castors have plated horns and axles