Tisk…
Single-sided step ladders
Dodavatel: GUENZBURGER STEIGTECHNIK
41103 41903 40103 41503 41504
139-0650EA 0 CZK
139-0650 139-0651 139-0769 GUEN41503 GUEN41504
Single-sided step ladders
Žebříky a schůdky
Ascendable on one side, grooved steps with a depth of 80 mm with durable 'step to side' rail connection, non slip nivello® ladder feet. High standing safety ensured by conical construction.
  • Connection on both sides by two detachable metal struts
  • Spacing between steps: 235 mm
  • Ladder inclination: 20°
  • Non slip nivello® ladder shoes
  • Ergopad grip zone
  • Safe cap joint
  • Durable 'step to side' rail connection
Certifikace: GS tested; in accordance with European standard DIN EN 14183.
Action manual BGI 694 and the applicable accident prevention regulations.