Tisk…
Transport trolleys for lab
Dodavatel: Metro International
CHEMX3 LABX3 MDSX3
139-0059EA 44380 CZK
139-0059 139-0075 139-0076
Transport trolleys for lab
Vozíky
Metro’s Lab Cart line includes a General Lab Cart, Chemical Cart, and Mobile Desk. All carts are designed to offer superior corrosion resistance. Smooth rolling, 127 mm polyurethane castors come as standard with each cart. All shelves adjust in 12,7 mm increments, offering precise shelf adjustment and maximum storage utilisation. Microban antimicrobial protects the lower shelf mat from bacteria, mold, mildew and fungi that cause odors, stains and product degradation.
Mobile Desk:
Mobile Desk can be used for instrumentation storage or can be combined with accessories to provide a functionalMore Product Information