Tisk…

Arzen, 1000 mg/L in 5% kyselině dusičné (from As) Specpure® standard pro ICP

Dodavatel: Thermo Fisher Scientific

13836.AD 13836.AE 13836.AP
13836.ADEA 1850 CZK
13836.AD 13836.AE 13836.AP
Arzen, 1000 mg/L in 5% kyselině dusičné (from As) Specpure® standard pro ICP
Standardy Standardy kationické Standrady - arzen

Molecular Formula: As in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309


Certifikace: Traceable to NIST

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR