Tisk…

Zinek 1000 mg/L Zn in 5% kyselině dusičné (from Zn) Specpure® standard pro ICP

Dodavatel: Thermo Fisher Scientific

13835.AD 13835.AE 13835.AP
13835.ADEA 1820 CZK
13835.AD 13835.AE 13835.AP
Zinek 1000 mg/L Zn in 5% kyselině dusičné (from Zn) Specpure® standard pro ICP
Standardy Standardy kationické Standardy - zinek

Molecular Formula: Zn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011291


Certifikace: Traceable to NIST

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR