Tisk…

Hydrogensíran sodný monohydrát, Reagent Grade

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Sodium bisulphate monohydrate

13799.36 13799.A1 13799.A7
13799.36EA 2260 CZK
13799.36 13799.A1 13799.A7
Hydrogensíran sodný monohydrát, Reagent Grade
Hydrogensíran sodný monohydrát

CAS No.: 10034-88-5 Molecular Formula: NaHSO4.H2O Formula Weight: 138.07 (120.06anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Colourless Melting Point: ca 183° dec. Merck Reference: 14,8586 UN No.: 3260 MDL No.: MFCD00149210

Vzorec: NaHSO₄·H₂O
MW: 138.08
Bod tání: ∼183 °C (rozkl.)
Hustota: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS číslo: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8 III
Merck Index: 14,8586

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR