Tisk…
Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Dodavatel: DUPERTHAL
29-200667-030 29-200967-030 29-201267-030 29-201267-032 29-200667-030L 29-200667-430L 29-130667-430 29-131267-030 29-130667-030 29-130667-030L 29-201267-050 29-201267-051 29-201267-052 29-201267-053 29-201267-056 29-201267-057 29-201267-058 29-201267-059 29-200967-050 29-200967-051 29-200967-052 29-200967-053 29-200677-053 29-200667-050 29-200667-051 29-200667-052 29-200667-053 29-200677-053L 29-200667-050L 29-200667-051L 29-200667-052L 29-200667-053L 29-201267-430 29-201267-432 29-200967-430 29-200667-430 29-130667-430L 29-131267-430 29-201267-450 29-201267-451 29-201267-452 29-201267-453 29-201267-459 29-200967-450 29-200967-451 29-200967-452 29-200967-453 29-200667-451 29-200667-452 29-200667-453 29-200677-453L 29-200667-450L 29-200667-451L 29-200667-452L 29-200667-453L 29-201265-030
135-1958EA 68630 CZK
135-1958 135-1685 132-1308 132-1309 135-2501 135-2511 132-1304 132-1306 132-1302 132-1303 132-1310 132-1312 132-1313 132-1314 132-1315 132-1316 132-1317 132-1318 135-2524 135-2525 135-2526 135-2527 135-2520 135-2502 135-2504 135-2506 135-2508 135-2521 135-2503 135-2505 135-2507 135-2509 132-1319 132-1320 135-2528 135-2510 132-1305 132-1307 132-1321 132-1322 132-1323 132-1324 132-1328 135-2529 135-2530 135-2531 135-2532 135-2514 135-2516 135-2518 135-2523 135-2513 135-2515 135-2517 135-2519 DUEH29-201265-030
Safety cabinets, type 90, with wing door technology, CLASSIC line standard and CLASSIC line pro
Nábytek kancelářský Skříně pro zásoby
Safety cabinets with storage shelves (CLASSIC line standard) or pull-out shelves (CLASSIC line pro) for the storage of hazardous substances in working spaces. Complete carcass, interior fittings and wing doors are made of powder-coated sheet steel.
  • Doors with standard integrated arrest system, meaning that containers can be placed into and removed from the cabinet with both hands
  • Removable base and adjustable feet for easy alignment and transport with a hand pallet truck.
  • The base is set back to prevent hitting one’s feet
  • Ventilation at every cabinet
More Product Information