Tisk…
Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)
Dodavatel: CleanAir by Baker
NH611003 NH411003 NH411001 NH611001
135-1871EA 639810 CZK
135-1871 135-1867 135-1865 135-1869
Biological safety cabinet, BioVanguard B, for cytotoxic (high risk) applications (cytotoxic medicines)
Digestoře Biologické boxy
BioVanguard Greenline B is a class II cabinet range which offers simplicity, robustness and high reliability ensuring the highest level of protection for operator, product and the environment, minimising hazards inherent to working with agents assigned to biosafety levels 1, 2 and 3. BioVanguard B is designed for high risk microbiological and high toxic applications, such as the production of cytotoxic medicines.
  • Easy to install, easy to operate, easy to clean, easy to decontaminate with low maintenance
  • Microprocessor control with automatic filter clogging
More Product Information