Tisk…
Under bench cabinets, type 90, UB-line
Dodavatel: asecos
30401001 30402001 30420001 30421001 30427001 30428001 30457001 30458001 30465001 30472001 30473001 32657001 32658001
135-2026EA 85790 CZK
135-2026 135-2027 135-1862 MSGA30421001 MSGA30427001 MSGA30428001 135-2622 MSGA30458001 MSGA30465001 135-2030 MSGA30473001 MSGA32657001 MSGA32658001
Under bench cabinets, type 90, UB-line
Nábytek kancelářský Skříně pro zásoby
Under bench cabinets with drawers or wing doors. Fire resistance: 90 minutes.
  • Safety - right drawer standard fitted with earthing wire incl. clamp, both drawer with seal welded sump and self-closing in the event of fire
  • Drawers are lockable with cylinder lock and remain open in any position
  • Flexible and mobile – castors with plinth optionally available
  • Integrated air ducts ready for connection (NW 75) to a technical exhaust system technical ventilation of under bench cabinets: recirculating air filter system UFA.20.30-AUS
EN 14727 conformity (More Product Information