Tisk…
Skříň na plynové láhve, typ 30, G-CLASSIC-30
Dodavatel: asecos
30651001 30663001 30673001 30685001 30686001
135-1826EA 114440 CZK
135-1826 135-1829 135-1832 135-1835 135-1837
Skříň na plynové láhve, typ 30, G-CLASSIC-30
Nábytek kancelářský Skříně pro zásoby
Fire-resistant type 30 safety cabinets for the safe storage of gas cylinders in buildings, in accordance with DIN EN 14470-2.
  • Easy installation of equipment and pipes - large interior height (1890 mm) and adjusting aids to compensate for uneven floor
  • Access possibilities on the top of the cabinet
  • No unauthorised use - doors lockable with cylinder lock (suitable for locking systems)
  • Door opening angle max. 180°
  • Quickly put into operation - integrated air ducts ready for connection to a technical exhaust system, complete and uniform ventilation of the
More Product Information