Tisk…
Safety storage cabinets, type 90, Q-PEGASUS-90
Dodavatel: asecos
30002040 30002041 30002046 30002047 30002048 30002049 30002050 30042040 30042041 30042046 30042047 30042048 30042049 30042050 30074040 30074041 30074046 30074047 30074048 30074049 30074050 30075040 30075041 30075046 30075047 30075048 30075049 30075050
135-1743EA 119180 CZK
135-1743 135-1744 135-2358 135-2359 135-2360 135-2361 135-2362 135-1750 135-1751 135-2371 135-2372 135-2373 135-2374 135-2375 135-1754 135-1755 135-2400 135-2401 135-2402 135-2403 135-2404 135-2405 135-2406 135-2407 135-2408 135-2409 135-2410 135-2411
Safety storage cabinets, type 90, Q-PEGASUS-90
Nábytek kancelářský Skříně pro zásoby
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient with transport base and bicoloured cabinet design. The PEGASUS line is nearly identical to the CLASSIC line, but with integrated AGT (one handed operation for double door cabinets) and integrated TSA (door open arrest system and automatic door closing).
  • Robust construction and longevity - triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch and impact resistant
More Product Information