Tisk…
Microbiological safety cabinets, Class II, EF and EF/B
Dodavatel: CleanAir by Baker
NP0301104ARN NP1991104 NP1991105 NP1991106 NP1992104 NP1992106 NP0302104RN NP0302106RN NP0301105ARN
135-0719EA 0 CZK
135-0719 135-0058 135-0059 135-0060 135-0104 135-0105 135-1499 135-0723 135-0720
Microbiological safety cabinets, Class II, EF and EF/B
Digestoře Biologické boxy
Microbiological safety cabinets provide maximum safety for the user, product and environment. Suitable for all applications in the fields of molecular biology, biotechnology, pharmaceuticals and medical research.
  • The window can be hermetically sealed, which is very important for gas disinfection
  • All models feature epoxy coated exterior and stainless steel interior
  • Conveniently designed for maintenance, the HEPA filter is replaced simply by removing it from the front panel
  • Alarm function (wrong air velocity or incorrect window position) directly in the user'