Tisk…
VWR® Star Tec, ESD Chairs
Dodavatel: VWR Collection
132-1396EA 12200 CZK
132-1396 132-1397 132-1398 132-1399 132-1400 132-1401
VWR® Star Tec, ESD Chairs
Židle
ESD chairs, with conductive seat and back, in black artificial leather, are soft and comfortable. The permanent backrest follows all movements and supports dynamic sitting in laboratories. Extra large, ergonomically-designed backrest with integrated lumbar support, the upholstery pads on the backrest provide maximum seating comfort and an optimum micro climate. The tapered backrest, in the upper area, offers elbow room for flexible work in laboratories and maximum freedom of movement.
  • Reliable ESD protection
  • Washable, resistant to desinfectants
  • Robust and
More Product Information