Tisk…

Lanthanum(III) bromide hydrate ≥99.99%

Dodavatel: Thermo Fisher Scientific
Varování

13198.18 13198.30 13198.09
13198.18EA 4330 CZK
13198.18 13198.30 13198.09
Lanthanum(III) bromide hydrate ≥99.99%
Lanthanum(III) bromide hydrate

Purity: 99.99% CAS No.: 224183-16-8 Molecular Formula: LaBr3.xH2O Formula Weight: 378.62(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White MDL No.: MFCD00798151

Vzorec: Br₃H₂LaO
Storage Temperature: Ambient
MDL Number: MFCD00798151
CAS číslo: 224183-16-8
EINECS: 236-896-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR