Tisk…

Nickel(II) telluride ≥99.9% (metals basis excluding Co) (max. 0.1-1% Co)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

13105.09 13105.18
13105.09EA 2700 CZK
13105.09 13105.18
Nickel(II) telluride ≥99.9% (metals basis excluding Co) (max. 0.1-1% Co)
Nickel(II) telluride

Purity: 99.9% CAS No.: 12142-88-0 Molecular Formula: NiTe Formula Weight: 186.31 Physical Form: 10mm & Down Lump Odour: Odourless UN No.: 3077 MDL No.: MFCD00016258

Vzorec: NiTe
Storage Temperature: Ambient
MDL Number: MFCD00016258
CAS číslo: 12142-88-0
EINECS: 235-260-6
UN: 3077
ADR: 9,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR