Tisk…
Autoclavable muff covers with skirt, Tyvek®
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
MFT2A05SK02 MFT2A07SK02 MFT2A09SK02 MFT2A12SK02 MFT2A18SK02 MFT2A18SK02TL
129-3124EA 180870 CZK
129-3124 129-3127 129-3130 129-3132 129-3135 129-3136
Autoclavable muff covers with skirt, Tyvek®
Návleky ochranné
Save time and standardize the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic and lint-free Tyvek® 1422A
  • With elastic to fit a range of diameters
  • With skirt extension for easy removal
  • Conveniently packaged 100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization.More Product Information