Tisk…
Autoclavable muff covers, with Tyvek® drawstring, BHD
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
MFNWB05D MFNWB07D MFNWB09D MFNWB12D MFNWB16D
129-3060EA 147260 CZK
129-3060 129-3061 129-3063 129-3065 129-3066
Autoclavable muff covers, with Tyvek® drawstring, BHD
Návleky ochranné
Save time and standardise the activities in equipment preparation for sterilisation.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear- and puncture-resistant
  • Significant process improvement compared to wrapping with sheets and tape
  • Tyvek® drawstring closure
BHD Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can beMore Product Information