Tisk…
BHD Autoclavable Stopper Bowl Covers
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
BWLNWB24 BWLNWB30 BWLNWB36 BWL2NWB36 BWLNWB48 BWL2NWB48 BWLNWB52 BWLNWB36-2PLY8 BWLNWB48NTTNSD
129-2794EA 121380 CZK
129-2794 129-2795 129-2868 129-2792 129-2869 129-2793 129-2870 KACCBWLNWB36-2PLY8 KACCBWLNWB48NTTNSD
BHD Autoclavable Stopper Bowl Covers
Návleky ochranné
Save time and standardise the activities in equipment preparation for sterilisation. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination, and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • With elastic to fit a range of diameters
  • Tear- and puncture-resistant blue BHD
More Product Information