Tisk…
Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Dodavatel: KEYSTONE CLEANROOM PRODUCTS
BNWBG0606D2P BNWBG0642D2P BNWBG09.512D2P BNWBG1212D2P BNWBG1218D2P BNWBG1624D2P BNWBG01644D2P BNWBG1830D2P BNWBG1854D2P BNWBG2024D2P BNWBG2032D2P BNWBG2424D2P BNWBG2435D2P BNWBG2436D2P BNWBG3648D2P BNWBG4040D2P BNWT3654D22P
129-2776EA 190470 CZK
129-2776 129-2777 129-2778 129-2779 129-2780 129-2781 129-2770 129-2782 129-2783 129-2784 129-2785 129-2786 129-2787 129-2788 129-2789 129-2790 129-2791
Autoclave Bags with Steam Indicator and Tyvek® Drawstring Closure, BHD
Sáčky Obaly sterilizační
BHD autoclave bags with Tyvek® drawstrings can be used as either the inner, or outer bag for the steam sterilisation of pharmaceutical change parts.
  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Simplifies conformance when wrapping to SOP's
  • Steam sterilisation indicator - autoclavable up to 135 °C
  • Tear and puncture resistant 2 ply blue BHD non woven
  • Significant process improvement compared to wrapping with sheets and tape
Standardise and simplify the way equipment is wrapped for steam sterilisation. The use ofMore Product Information