Tisk…
BRUTE® Square Containers
Dodavatel: Rubbermaid Commercial Products
FG352600GRAY FG352600RED FG353600GRAY
129-1789EA 4510 CZK
129-1789 129-1788 129-1786
BRUTE® Square Containers
Koše odpadkové a zásobní Koše odpadní
BRUTE® square containers with increased capacity are suitable for storage or refuse collection.
  • Square shape offers up to 14% more capacity than round containers
  • Nesting handles allow secure fit for side-by-side use
  • Rounded corners and smooth contours are easy to clean
  • All BRUTE® square containers come with a 10-year warranty
Certifikace: Certified EU food contact compliant (grey only). Meets NSF 2, 21 approval and USDA meat and poultry group listed, ensuring regulatory compliance for food storage and clean ability.
Informace o príslušenství: BRUTE®More Product Information