Tisk…
Sustainable 2-in-1 flat mop covers, Revolve™
Dodavatel: Texwipe
TX1720 TX1721
129-1777EA 23630 CZK
129-1777 129-1778
Sustainable 2-in-1 flat mop covers, Revolve™
Mopy
Revolve™ 2-in-1 mop covers, the first sustainable covers made for the cleanroom industry, are made from 100% upcycled polyester. Revolve™ mop covers contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Mop and clean large and small surfaces
  • Apply and remove disinfectants
  • Spill control
  • 100% upcycled polyester
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
  • Appropriate for use with temperatures less than 400 °F (205 °C)
More Product Information
Objednat
Produktová specifikace
Environmentally Preferable Waste Reducing,Udržitelné materiály