Tisk…
Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Dodavatel: Bel-Art Products, a Part of SP
H131931000 F131930102 F131930500
129-4948EA 1650 CZK
129-4948 129-1475 129-1654
Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Stojany Držáky sáčků
Economical and convenient, this bench top disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc during daily use.
  • Snug fitting polypropylene plastic cover helps hold the bag securely in place
  • Reusable, can be cleaned by hand or in automated dish washers
  • Autoclavable
The rectangular steel wire frame holds a 220×280 mm polyethylene bag which is coated in easy to clean, durable, orange colour epoxy.
Certifikace: Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used toMore Product Information