Tisk…
VWR®, Biodegradable Bags, Twist-Seal
Dodavatel: VWR Collection
129-1488EA 11010 CZK
129-1488 129-1516 129-1538 129-1541 129-1543 129-1546
VWR®, Biodegradable Bags, Twist-Seal
Sáčky Sáčky na vzorky
LDPE bags with safety tabs, manufactured and packed in sterile conditions.
  • Provides a secure, contaminant-free, flexible container that ensures dependandle analytical results
  • RNase-, DNase-, pyrogen- and BPA-free
  • Offers an economical and efficient way to collect,contain and carry samples
  • Wire tab keeps bag open for filling
  • Unique double fold over system provides a temporary leakproof seal
Made of highly resistant virgin LDPE containing additives that promote in landfill conditions. The biodegradation does not start before it is put in landfillMore Product Information