Tisk…
Mops, MegaSwat™
Dodavatel: MICRONOVA
MS6-16NS MS6-8 MS6-8IR MSAA-16NS MS6-16NSIR MS1111-16 MS6-8NSIR MS6-18NSIR MS6-18NS
129-0518EA 660 CZK
129-0518 129-0614 129-1083 MICRMSAA-16NS MICRMS6-16NSIR MICRMS1111-16 MICRMS6-8NSIR MICRMS6-18NSIR MICRMS6-18NS
Mops, MegaSwat™
Mopy
These flat, paddle-shaped mops are for sliding behind heavy cabinets and process equipment, over piping and underneath calibrated equipment. Mop heads measure (W×D) 406×3 mm.
  • Flexible polyethylene interior, choice of PVA, NovaLite latex rayon or NovaPoly polyester covering
  • MegaSwat™ Lite with NovaLite cover withstands stronger disinfectants and can be autoclaved
  • Mop heads attach to universal stainless steel handle via quick disconnect MegaSwat™ adapter
Informace pro objednávání: All components must be ordered separately.
Objednat
Produktová specifikace
Environmentally Preferable Waste Reducing