Tisk…
Mop na podlahy bez kbelíku, EasySat™
Dodavatel: CONTEC
BLVKMOP BLVKMOPIR TBLVKMOP BLMOPQC
129-0213EA 14650 CZK
129-0213 129-0214 129-0215 141-1467
Mop na podlahy bez kbelíku, EasySat™
Mopy
EasySat™ bucketless floor mop is an effective and quick solution to cleaning challenges.
  • Fully autoclavable
  • Closed solution tank minimises exposure to chemicals
  • Large opening makes tank easy to fill
  • Lightweight mop system weighs 3,2 kg with full tank
  • Trigger flow control mechanism releases cleaning solution on demand or continuously
  • Angled mop head cleans hard-to-reach areas and is available in two styles: smooth surface for standard floor cleaning, and textured surface for more abrasive action
Designed for the cleanroom, this innovative mop is easyMore Product Information