Tisk…

hořčík, ribbon, Width: 3.0 mm (0.12 in), Thickness: 0.15 mm (0.0059 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

12811.15 12811.A0 12811.14 12811.51
12811.15EA 0 CZK
12811.15 12811.A0 12811.14 12811.51
hořčík, ribbon, Width: 3.0 mm (0.12 in), Thickness: 0.15 mm (0.0059 in)
hořčík

CAS No.: 7439-95-4 Physical Form: ≈0.78g/m Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Vzorec: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS číslo: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR