Tisk…

Potassium tetrabromoaurate(III) dihydrate (≥33% Au) ≥99.99% (metals basis), Premion®

Dodavatel: Thermo Fisher Scientific
Varování

12664.03 12664.06
12664.03EA 3810 CZK
12664.03 12664.06
Potassium tetrabromoaurate(III) dihydrate (≥33% Au) ≥99.99% (metals basis), Premion®
Potassium tetrabromoaurate(III) dihydrate

Purity: 99.99% CAS No.: 14323-32-1 Molecular Formula: KAuBr4.2H2O Formula Weight: 591.72 (555.69anhy) Physical Form: Crystalline Odour: Weak Solubility: Soluble in water and alcohol MDL No.: MFCD00049650

Vzorec: KAuBr₄.2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00049650
CAS číslo: 14323-32-1
EINECS: 238-268-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR