Tisk…

Hafnium disilicide

Dodavatel: Thermo Fisher Scientific

Synonyms: Hafnium silicide

12564.09
12564.09EA 5670 CZK
12564.09
Hafnium disilicide
Hafnium disilicide

CAS No.: 12401-56-8 Molecular Formula: HfSi2 Formula Weight: 234.66 Physical Form: Powder Odour: Odourless Melting Point: 1680° MDL No.: MFCD00167026

Vzorec: HfSi₂
MW: 234.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00167026
CAS číslo: 12401-56-8
EINECS: 235-640-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR