Tisk…

Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Packaged under argon.

12396.FF 12396.FI
12396.FFEA 4290 CZK
12396.FF 12396.FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈0.49g/25x25mm, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver UN No.: 3178 MDL No.: MFCD00011022

Vzorec: Gd
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS číslo: 7440-54-2
EINECS: 231-162-2
UN: 3178
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR