Tisk…

Tellurium tetrachloride ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Tellurium(IV) chloride

12284.14 12284.22
12284.14EA 4880 CZK
12284.14 12284.22
Tellurium tetrachloride ≥99.9% (metals basis)
Tellurium tetrachloride

Purity: 99.9% CAS No.: 10026-07-0 Molecular Formula: Cl4Te Formula Weight: 269.41 Physical Form: Crystalline Density: 3.01 Appearance: Yellow Odour: Pungent Melting Point: 224° Boiling Point: 380° Solubility: Decomposes in water forming TeO2 and HCl Merck Reference: 14,9126 UN No.: 3260 MDL No.: MFCD00011262

Vzorec: Cl₄Te
MW: 269.41
Bod varu: 380 °C
Bod tání: 224 °C
Hustota: 3.01
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS číslo: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8 II
Merck Index: 14,9126

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR