Tisk…

Sodium molybdate(VI) Mo 46,2%, anhydrous

Dodavatel: Thermo Fisher Scientific
Varování

12209.22 12209.36
12209.22EA 1780 CZK
12209.22 12209.36
Sodium molybdate(VI) Mo 46,2%, anhydrous
Sodium molybdate(VI)

CAS No.: 7631-95-0 Molecular Formula: MoNa2O4 Formula Weight: 205.92 Physical Form: Powder Density: 3.72 Refractive Index: 1.714 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 687° Merck Reference: 14,8645 MDL No.: MFCD00003486

Vzorec: MoNa₂O₄
Bod tání: 687 °C
Hustota: 3,72
Storage Temperature: Ambient
MDL Number: MFCD00003486
CAS číslo: 7631-95-0
EINECS: 231-551-7
Merck Index: 14,8645

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR