Tisk…
PIG® leak diverters for roofs
Dodavatel: NEW PIG
TLS462-TR TLS463-TR TLS464-TR TLS465-TR TLS466-TR TLS467-TR TLS552-TR TLS553-TR TLS554-TR TLS462-CL
121-0159EA 2560 CZK
121-0159 121-0160 121-0161 121-0162 121-0163 121-0164 121-0168 121-0169 121-0170 NEWPTLS462-CL
PIG® leak diverters for roofs
Absorpční materiály
Leak diverters help protect people, equipment and inventory from a roof leak.
  • Hangs easily in problem spots to catch nuisance leaks and channel them to a floor drain or collection container
  • Ideal for quick response to avoid downtime after a roof leak or for long-term prevention in leak-prone areas
  • Nonpermeable tarp is vinyl coated for strength and durability; resists punctures, UV and mildew for long life
  • Pinched corners help to funnel liquid toward centre for better drainage
Accepts a standard 3/4" hose connection to channel liquid to floor drain orMore Product Information