Tisk…
PIG® Original DrainBlocker® drain covers
Dodavatel: NEW PIG
PLRE241 PLRE244 PLRE242 PLRE243 PLRE245
121-0048EA 4280 CZK
121-0048 121-0049 121-0050 NEWPPLRE243 NEWPPLRE245
PIG® Original DrainBlocker® drain covers
Absorpční materiály
PIG® DrainBlocker® Covers deploy quickly and create a tight, temporary seal over drains to keep out unwanted liquids.
  • Exclusive New Pig formulation features a super-durable, tear-resistant top layer that withstands repeated use and abrasion combined with a flexible, tacky bottom layer that tightly seals drain openings
  • Chemical-resistant polyurethane resists water, oil and many chemicals
  • Flexible Cover moulds to uneven surfaces to create an instant, temporary drain seal that prevents pollution
  • High-visibility yellow top colour draws attention to spill for
More Product Information