Tisk…

Rhodium ≥99.95% (metals basis), powder -60 mesh

Dodavatel: Thermo Fisher Scientific
Varování

12072.02 12072.03 12072.06
12072.02EA 42250 CZK
12072.02 12072.03 12072.06
Rhodium ≥99.95% (metals basis), powder -60 mesh
Rhodium

Purity: 99.95% CAS No.: 7440-16-6 Appearance: Grey Odour: Odourless UN No.: 3089 MDL No.: MFCD00011201

Vzorec: Rh
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS číslo: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR