Tisk…

Olovo 99,998 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

12049.FF 12049.FI
12049.FFEA 3190 CZK
12049.FF 12049.FI
Olovo 99,998 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Olovo

Purity: 99.998% CAS No.: 7439-92-1 Physical Form: ≈3.5g/25x25mm Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Vzorec: Pb
MW: 207.2
Bod varu: 1740
Bod tání: 327
Hustota: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS číslo: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR