Tisk…
Dispensing bottles for flammable liquids
Dodavatel: JUSTRITE MANUFACTURING
14011 14009
119-0276EA 1260 CZK
119-0276 119-0274
Dispensing bottles for flammable liquids
Nádoby bezpečnostní
PE bottles with stainless steel wire.
  • Provide safe dispensing of flammable liquids directly on small components or subassemblies with a minimum of over-spray
  • Squeezing the bottle produces a fine stream of liquid
  • Stem can be shaped to dispense into hard-to-get areas