Tisk…
Eyewash station, QuickSafe® Basic
Dodavatel: Plum Safety
5170
118-0398EA 3440 CZK
118-0398
Eyewash station, QuickSafe® Basic
Pomůcky první pomoci Sprchy na výplach očí
This wall-mountable station is made of impact-resistant plastic and equipped with a transparent cover.
  • Users can tailor the first shelf with products that fit their business
  • Cabinet is only slightly larger than an A3 sheet of paper
  • Comes with instructions and fixtures for wall mounting
Content:
2×500 ml Plum Eye Wash
1×200 ml pH Neutral
5×QuickRinse eyewash ampoules (20 ml)
1×QuickFix Elastic plaster refill
W×D×H: 253×92×430 mm