Tisk…

Rhodium ≥99.8% (metals basis), wire, Ø 0.5 mm (0.02 in)

Dodavatel: Thermo Fisher Scientific

11576.BSEA 21520 CZK
11576.BS 11576.BR 11576.BV 11576.CB
Rhodium ≥99.8% (metals basis), wire, Ø 0.5 mm (0.02 in)
Rhodium
Vzorec: Rh
Bod varu: 3400…3700 °C (1013 hPa)
Bod tání: 1960 °C
Hustota: 12,41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011201
CAS číslo: 7440-16-6
EINECS: 231-125-0
Merck Index: 12,08353

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR