Tisk…

Oxid zinečnatý ≥99.0% ACS

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Zinci oxidum, Zinc (II) oxide

11558.36 11558.A3
11558.36EA 2300 CZK
11558.36 11558.A3
Oxid zinečnatý ≥99.0% ACS
Zinc oxide

Purity: 99.0% min CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: -325 Mesh Powder Density: 5.606 Melting Point: 1975° Solubility: Insoluble in water. Soluble in dilute acetic acid, dilute mineral acids, ammonia, ammonium carbonate Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300


Pigment in white paints, UV absorber in plastics, ceramics, piezoelectric devices

Vzorec: ZnO
Bod tání: 1975 °C
Hustota: 5,606
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS číslo: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR