Tisk…
Wipes for general applications, Multizell®
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
10440B 10440A 10333B 10333A
115-2761EA 2640 CZK
115-2761 115-2762 115-2763 115-2764
Wipes for general applications, Multizell®
Ubousky/Utěrky Utěrky čistící pro opakovatelné použití Utěrky pro běžné aplikace
These wipes are made from 100% recycled material (cellulose). Ideal to wipe off liquids or to remove soiling. Wipes can be taken one at a time, available in 3 or 4 ply versions and in various sizes.
  • Highly absorbent
  • Lint-free
  • Not bleached