Tisk…
Wipe rolls, Multisoft® Air
Dodavatel: Zellstoff-Vertriebs-GmbH & Co. KG
11781
115-2728EA 3220 CZK
115-2728
Wipe rolls, Multisoft® Air
Ubousky/Utěrky Utěrky jednorázové a ručníky Ubrousky pro běžné aplikace
High quality wipes for cleaning and polishing sensitive surfaces such as metal, plastics, wood, or finishes. Multi-soft cotton fleece roll with 500 sheets.
  • Extremely soft, highly absorbent and solvent-resistant
  • Strong, low-linting with special embossment
Colour: White
Colour: White