Tisk…
Revolve™ Sustainable Cleanroom Wipes
Dodavatel: Texwipe
TX1704 TX1709 TX1712
115-2714EA 25940 CZK
115-2714 115-2718 115-2730
Revolve™ Sustainable Cleanroom Wipes
Ubousky/Utěrky Utěrky pro čisté prostory Ubrousky nevlhčené do čistých prostor
Revolve™ wipes, sustainable wipes made for the cleanroom industry, are made from 100% upcycled polyester material with a sealed edge, processed on Texwipe’s fully automated manufacturing system. Revolve™ wipes contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Wipe and clean surfaces, equipment, and parts
  • Apply and remove lubricants, adhesives, residues, and other solutions including disinfectants
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol,
More Product Information